HLA microtest plates (Terasaki plates)
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
653190_480 653180_270
391-3611EA 498 EUR
391-3611 391-3612
HLA microtest plates (Terasaki plates)
Microplaten Immunologie-microplaten en -strips
PS, cell culture treated surface, with lid. HLA microtest plates (Terasaki plates) are suitable for all applications requiring the serological determination of HLA antigens.
  • Choice of 60- or 72-well plates
  • 10 µl volume per well