VWR®, Cell culture media, Dulbecco's Modified Eagles Medium (DMEM)
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
392-0407EA 8.8 EUR
392-0407 392-0408 392-0409 392-0410 392-0411 392-0412 392-0413 392-0414 392-0415 392-0416 392-0417
VWR®, Cell culture media, Dulbecco's Modified Eagles Medium (DMEM)
Celcultuurmedia
Dulbecco's Modified Eagle Medium (DMEM) is designed to preserve and maintain the growth of a broad spectrum of mammalian cell types.
  • Suitable for most cell types
  • Available in a broad variety of formulations
  • Also available with stable glutamine (L-Alanyl-L-Glutamine) for a better shelf life
The DMEM low glucose has been developed for the culture of mouse embryonic cells.