Formaline 10% v/v, histology grade, neutralized (pH 7.0 ± 0.2), J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Formaldehyde

3933.9020EA 36.49 EUR
3933.9020 3933.5000PC 3933.9010 3933.1000 3933.1000MB 3933.9010JL 3933.9020JL
Formaline 10% v/v, histology grade, neutralized (pH 7.0 ± 0.2), J.T.Baker®
Formalin
Formule: CH₂O
MW: 30.03 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003274
CAS nummer: 50-00-0
EINECS: 200-001-8
IVDLogo

Order Now

Specification Test Results

Natrium (Na) 61 - 71 mmol/l
Formaldehyde (HCHO) 38 - 42 g/l
pH bij 25°C 6,8 - 7,2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR