Sodium thiocyanate ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Sodium rhodanide

3938-07EA 0 EUR
3938-07
Sodium thiocyanate ≥98.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium thiocyanate
Formule: NaSCN
MW: 81.07 g/mol
Smeltpunt: 287
Dichtheid: 1.735 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011123
CAS nummer: 540-72-7
EINECS: 208-754-4
Merck Index: 13,09401

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements = 98.0 %
For Laboratory,Research,or Manufacturing Use = 98.0 %
ACS - Assay (NaSCN) ≥ 98.0 %
ACS - Insoluble Matter ≤ 0.005 %
ACS - Carbonate (as Na₂CO₃) ≤ 0.2 %
ACS - Chloride (Cl) ≤ 0.01 %
ACS - Sulfate (SO₄) ≤ 0.01 %
ACS - Sulfide (S) ≤ 0.001 %
ACS - Ammonium (NH₄) ≤ 0.002 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 2.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR