1-(Trimethylsilyl)imidazole, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

394874-10X1ML 394874-25ML 394874-5ML
394874-10X1MLEA 285 EUR
394874-10X1ML 394874-25ML 394874-5ML
1-(Trimethylsilyl)imidazole, Supelco®
1-(Trimethylsilyl)imidazole
Formule: C₆H₁₂N₂Si
Kookpunt: 209 °C (1013 hPa)
Smeltpunt: –42 °C
Dichtheid: 0,95 g/cm³ (20 °C)
Vlampunt: 61 °C
MDL Nummer: MFCD00005280
CAS nummer: 18156-74-6
EINECS: 242-040-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b21350c4c0c7ad81fa62f67c6b5196ea1a77e34&vid=8070ee5278cbf346cffa2fa72e144a41f13f6c02&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR