Strontium aluminate ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

39647.14 39647.22
39647.14EA 114 EUR
39647.14 39647.22
Strontium aluminate ≥99.5% (metals basis)
Strontium aluminate

Purity: 99.5% CAS No.: 12004-37-4 Molecular Formula: SrAl2O4 Formula Weight: 205.58 Physical Form: -100 Mesh Powder Density: 3.559 Appearance: Silver Odour: Odourless MDL No.: MFCD00054054

Formule: Al₂O₄Sr
Dichtheid: 3,559
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054054
CAS nummer: 12004-37-4
EINECS: 234-455-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR