Sodium iodate ≥99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

40135.22 40135.36
40135.22EA 56.6 EUR
40135.22 40135.36
Sodium iodate ≥99%
Sodium iodate

Purity: 99% min CAS No.: 7681-55-2 Molecular Formula: NaIO3 Formula Weight: 197.89 Physical Form: Crystalline powder Density: 4.28 Appearance: White Melting Point: dec. Merck Reference: 14,8630 UN No.: 1479 MDL No.: MFCD00003533

Formule: INaO₃
Smeltpunt: Dec.
Dichtheid: 4,28
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003533
CAS nummer: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
Merck Index: 14,8630
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR