Potassium hydrogen carbonate 99.7-100.5% (dried basis) ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Potassium bicarbonate

40195.22 40195.36
40195.22EA 21.3 EUR
40195.22 40195.36
Potassium hydrogen carbonate 99.7-100.5% (dried basis) ACS
Potassium hydrogen carbonate

Purity: 99.7-100.5% CAS No.: 298-14-6 Molecular Formula: KHCO3 Formula Weight: 100.12 Physical Form: Granular Density: 2.17 Solubility: Slightly soluble in water. Practically insoluble in alcohol Merck Reference: 14,7609 MDL No.: MFCD00011402

Formule: KHCO₃
MW: 100.12 g/mol
Smeltpunt: 292
Dichtheid: 2.17
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011402
CAS nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 14,7609

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR