Neodymium(III) oxide ≥99% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40252.14 40252.22 40252.36 40252.PG
40252.14EA 55.2 EUR
40252.14 40252.22 40252.36 40252.PG
Neodymium(III) oxide ≥99% (REO, rare earth oxide basis), REacton®
Neodymium(III) oxide

Purity: 99% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: Powder Density: 7.24 Melting Point: 2272° Solubility: Insoluble in water. Soluble in acid Merck Reference: 14,6450 MDL No.: MFCD00011134


ceramic capacitors, Colouring glass, carbon arc-light electrodes, Colour TV tubes, dehydrogenation catalyst

Formule: Nd₂O₃
Kookpunt: 3760 °C (1013 hPa)
Smeltpunt: 2272 °C
Dichtheid: 7,24
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011134
CAS nummer: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR