Yttrium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40299.DH 40299.DJ 40299.DM 40299.HF
40299.DHEA 260 EUR
40299.DH 40299.DJ 40299.DM 40299.HF
Yttrium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Yttrium

Purity: 99.9% CAS No.: 7440-65-5 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.14g/mm, Cast rod, diameter is approximate Odour: Odourless

Formule: Y
Kookpunt: 2927 °C (1013 hPa)
Smeltpunt: 1525 °C
Dichtheid: 4,472 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011468
CAS nummer: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR