2-Butanonoxim 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

403331000 403330250 403330010
403331000.EA 26.6 EUR
403331000. 403330250. 403330010.
2-Butanonoxim 99%
2-Butanonoxim
Formule: C₄H₉NO
Kookpunt: 72 °C (25 mmHg)
Smeltpunt: –30 °C
Dichtheid: 0,923 g/cm³
Vlampunt: 62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013935
CAS nummer: 96-29-7
EINECS: 202-496-6
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4440 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR