Nickel chromium alloy (80:20 w%) ≥99% (metals basis), draad, Ø 1.0 mm (0.04 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

40438.G9 40438.H4 40438.H9
40438.G9EA 71.2 EUR
40438.G9 40438.H4 40438.H9
Nickel chromium alloy (80:20 w%) ≥99% (metals basis), draad, Ø 1.0 mm (0.04 in)
Nickel chromium alloy (80:20 w%)

Purity: 99% CAS No.: 11106-97-1 Molecular Formula: Ni:Cr; 80:20 wt% Physical Form: ≈6.6g/m Odour: Odourless

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00673110
CAS nummer: 11106-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR