Gold ≥99.995% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40442.KG 40442.KJ
40442.KGEA 186 EUR
40442.KG 40442.KJ
Gold ≥99.995% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Gold

Purity: 99.995% CAS No.: 7440-57-5 Physical Form: ≈0.49g/slug Odour: Odourless MDL No.: MFCD00003436


Electronics, vapor deposition

Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Dichtheid: 19.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR