Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.25 mm (0.01 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

40603.FI 40603.GH 40603.FF
40603.FIEA 237 EUR
40603.FI 40603.GH 40603.FF
Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.25 mm (0.01 in)
Magnesium

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈0.27g/25x25mm, Temper: as rolled Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Formule: Mg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085308
CAS nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR