Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness: 0.0075 µm

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aluminum

40682.DJ 40682.HF
40682.DJEA 1493 EUR
40682.DJ 40682.HF
Aluminium ≥99.995% (metals basis), ultrathin foil, Thickness: 0.0075 µm
Aluminium

Purity: 99.995% CAS No.: 7429-90-5 Physical Form: Circular disc on permanent mylar support Odour: Odourless MDL No.: MFCD00134029


Microelectronics, contacts and coatings

Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR