Gold ≥99.9% (metals basis), thin foil, Thickness: 0.005 mm (0.0002 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40739.FF 40739.FI
40739.FFEA 504 EUR
40739.FF 40739.FI
Gold ≥99.9% (metals basis), thin foil, Thickness: 0.005 mm (0.0002 in)
Gold

Purity: 99.9% CAS No.: 7440-57-5 Physical Form: ≈0.06g/25x25mm, not certified pinhole free Odour: Odourless MDL No.: MFCD00003436

Formule: Au
Kookpunt: 2800 °C (1013 hPa)
Smeltpunt: 1063 °C
Dichtheid: 19,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR