Polypropyleenglycol, M.W. ∼ 3,000

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40814.18 40814.30
40814.18EA 34 EUR
40814.18 40814.30
Polypropyleenglycol, M.W. ∼ 3,000
Polypropyleenglycol

CAS No.: 25322-69-4 Molecular Formula: H[OCH(CH3)CH2]nOH Physical Form: Liquid MDL No.: MFCD00084446

Formule: H(OCH₂CH₂CH₂)nOH
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00084446
CAS nummer: 25322-69-4
EINECS: 500-039-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR