Carbon ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40830.KS
40830.KSEA 656 EUR
40830.KS
Carbon ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Carbon

Purity: 99.999% Physical Form: Hot Pressed MDL No.: MFCD00133992

Formule: C
Kookpunt: ∼4000 °C (1013 hPa)
Smeltpunt: 3727 °C
Dichtheid: 3,65 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-955-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR