Tin ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40899.KS
40899.KSEA 280 EUR
40899.KS
Tin ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 6.35 mm (0.250 in)
Tin

Purity: 99.995% CAS No.: 7440-31-5 Odour: Odourless MDL No.: MFCD00133862

Formule: Sn
Kookpunt: 2270 °C (1013 hPa)
Smeltpunt: 232 °C
Dichtheid: 7,31 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133862
CAS nummer: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR