Titanium ≥99.7% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

40902.KS
40902.KSEA 460 EUR
40902.KS
Titanium ≥99.7% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Titanium

Purity: 99.7% CAS No.: 7440-32-6 Odour: Odourless MDL No.: MFCD00011264


Magnetron sputtering source

Formule: Ti
Kookpunt: 3262 °C (1013 hPa)
Smeltpunt: 1660 °C
Dichtheid: 4,51 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011264
CAS nummer: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR