Hastelloy C® alloy, gauze woven from wire 20 mesh woven from 0.23mm (0.009in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

40938.FL 40938.GT 40938.HB
40938.FLEA 38.2 EUR
40938.FL 40938.GT 40938.HB
Hastelloy C® alloy, gauze woven from wire 20 mesh woven from 0.23mm (0.009in)
Hastelloy C® alloy

Physical Form: Wire Cloth MDL No.: MFCD00801065


Applications: High strength, corrosion resistant alloy

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR