Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.25 mm (0.01 in) Type 304

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

40945.BWEA 27 EUR
40945.BW 40945.BZ
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), wire, Ø 0.25 mm (0.01 in) Type 304
Stainless steel

Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Odour: Odourless MDL No.: MFCD00148499

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR