Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, draad, Ø 0.025 mm (0.001 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41038.G5 41038.G6
41038.G5EA 318 EUR
41038.G5 41038.G6
Tungsten (gold coated) ≥99.95% (metals basis) Au 99.99%, draad, Ø 0.025 mm (0.001 in)
Tungsten (gold coated)

Purity: 99.95% Physical Form: ≈0.005g/m (W only), 3-5 wt% Au plating Odour: Odourless MDL No.: MFCD00011461


Microelectronics, evaporation source, high temperature

Formule: W
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011461
CAS nummer: 7440-33-7
EINECS: 231-143-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR