Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41072.KS
41072.KSEA 513 EUR
41072.KS
Magnesium oxide ≥99.95% (metals basis excluding Ca), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Magnesium oxide

Purity: 99.95% Molecular Formula: MgO; Ca 750ppm max Formula Weight: 40.30 MDL No.: MFCD00011109


Magnetron sputtering source

Formule: MgO
Kookpunt: 3600 °C (1013 hPa)
Smeltpunt: 2800 °C
Dichtheid: 3,58 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011109
CAS nummer: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR