Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99%, medium molecular weight, hydrolysed

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: PVOH, PVA, Mowiol®

41242.14 41242.22 41242.36
41242.14EA 39.3 EUR
41242.14 41242.22 41242.36
Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99%, medium molecular weight, hydrolysed
Polyvinyl alcohol

Purity: 98-99% CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odour: Odourless MDL No.: MFCD00081922

Formule: [-CH₂CH(OH)-]n
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081922
CAS nummer: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR