Sulfur in isooctane standard solutions, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Sulfur in isooctane standard solutions, Specpure®
Standaarden Standaarden voor de petroleumindustrie

Physical Form: Liquid
Appearance: Colourless
Odour: Petrol-like
UN No.: UN1262
MDL No.: MFCD00804269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR