Yttrium(III) fluoride, anhydrous ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Yttrium trifluoride, Yttrium(III) fluoride

41424.09 41424.18 41424.30
41424.09EA 78.8 EUR
41424.09 41424.18 41424.30
Yttrium(III) fluoride, anhydrous ≥99.9% (metals basis)
Yttrium(III) fluoride

Purity: 99.9% CAS No.: 13709-49-4 Molecular Formula: YF3 Formula Weight: 145.90 Physical Form: 3-12mm Pieces Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049714


For vacuum deposition

Formule: F₃Y
Kookpunt: 2227 °C (1013 hPa)
Smeltpunt: 1387 °C
Dichtheid: 4,01 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049714
CAS nummer: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR