Antimoontrichloride ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Antimony trichloride

41537.14 41537.22
41537.14EA 63.1 EUR
41537.14 41537.22
Antimoontrichloride ≥99.9% (metals basis)
Antimoontrichloride

Purity: 99.9% CAS No.: 10025-91-9 Molecular Formula: Cl3Sb Formula Weight: 228.11 Physical Form: Crystalline Density: 3.14 Appearance: White Odour: Pungent Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formule: SbCl₃
MW: 228.12 g/mol
Kookpunt: 223.5 °C
Smeltpunt: 73.4 °C
Dichtheid: 3.14
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011212
CAS nummer: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 14,707

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR