Calcium hexaboride ≥99% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

41551.14 41551.22
41551.14EA 138 EUR
41551.14 41551.22
Calcium hexaboride ≥99% (metals basis)
Calcium hexaboride

Purity: 99+% CAS No.: 12007-99-7 Molecular Formula: CaB6 Formula Weight: 104.95 Physical Form: -20 Mesh Powder Density: 2.30 Melting Point: 2235° MDL No.: MFCD00148904

Formule: B₆Ca
Smeltpunt: 2235 °C
Dichtheid: 2,30
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00148904
CAS nummer: 12007-99-7
EINECS: 234-525-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR