Antimony(III) bromide ≥99.5% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Antimony tribromide

41574.09 41574.18
41574.09EA 60.2 EUR
41574.09 41574.18
Antimony(III) bromide ≥99.5% (metals basis)
Antimony(III) bromide

Purity: 99.5% CAS No.: 7789-61-9 Molecular Formula: SbBr3 Formula Weight: 361.48 Physical Form: -8 Mesh Density: 4.148 Refractive Index: 1.74 Sensitivity: Moisture Sensitive Melting Point: 96.6° Boiling Point: 280° Merck Reference: 14,706 UN No.: 3260 MDL No.: MFCD00016317

Formule: Br₃Sb
MW: 361.47 g/mol
Kookpunt: 280 °C
Smeltpunt: 96.6 °C
Dichtheid: 4.148
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00016317
CAS nummer: 7789-61-9
EINECS: 232-179-8
UN: 3260
ADR: 8 II
Merck Index: 14,706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR