Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.5 mm (0.02 in) Type 304

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41585.FF 41585.FI 41585.GH
41585.FFEA 35.2 EUR
41585.FF 41585.FI 41585.GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness: 0.5 mm (0.02 in) Type 304
Stainless steel

Molecular Formula: Fe:Cr:Ni; 70:19:11 wt% Odour: Odourless MDL No.: MFCD00148499

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR