Strontium titanate S.A. 12.5-25 m²/g

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41634.22 41634.36
41634.22EA 198 EUR
41634.22 41634.36
Strontium titanate S.A. 12.5-25 m²/g
Strontium titanate

CAS No.: 12060-59-2 Molecular Formula: SrTiO3 Formula Weight: 183.51 Physical Form: 0.05-0.15 Micron APS Powder Density: 4.810 Odour: Odourless MDL No.: MFCD00049554

Formule: O₃SrTi
Smeltpunt: 2060 °C
Dichtheid: 4,810
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049554
CAS nummer: 12060-59-2
EINECS: 235-044-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR