Bismuth ≥99.999% (metals basis), foil, Thickness: 1.0 mm (0.04 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41636.FF 41636.FI
41636.FFEA 498 EUR
41636.FF 41636.FI
Bismuth ≥99.999% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Bismuth

Purity: 99.999% CAS No.: 7440-69-9 Physical Form: ≈6.1g/25x25mm Odour: Odourless MDL No.: MFCD00134033

Formule: Bi
Kookpunt: 1490 °C (1013 hPa)
Smeltpunt: 271…272 °C
Dichtheid: 9,78 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134033
CAS nummer: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR