Tungsten ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41640.KS
41640.KSEA 354 EUR
41640.KS
Tungsten ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Tungsten

Purity: 99.95% CAS No.: 7440-33-7 MDL No.: MFCD00011461

Formule: W
Kookpunt: 5660 °C (1013 hPa)
Smeltpunt: 3410 °C
Dichtheid: 19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011461
CAS nummer: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR