Sodium acetate hydrate ≥99.9985% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41685.06 41685.14 41685.22
41685.06EA 60.8 EUR
41685.06 41685.14 41685.22
Sodium acetate hydrate ≥99.9985% (metals basis), Puratronic®
Sodium acetate hydrate

Purity: 99.9985% CAS No.: 41484-91-7 Molecular Formula: NaOOCCH3.xH2O (x~3) Formula Weight: 82.03(anhy) Physical Form: Powder Density: 1.45 Melting Point: 58° Merck Reference: 14,8571 MDL No.: MFCD00071557

Formule: H₃CCOONa·3H₂O
Smeltpunt: 58 °C
Dichtheid: 1,45
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00071557
CAS nummer: 41484-91-7
EINECS: 204-823-8
Merck Index: 14,8571

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR