Cadmium(II) nitrate tetrahydrate ≥99.9% (metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Cadmium nitrate tetrahydrate

41732.18 41732.30
41732.18EA 116 EUR
41732.18 41732.30
Cadmium(II) nitrate tetrahydrate ≥99.9% (metals basis)
Cadmium(II) nitrate tetrahydrate

Purity: 99.9% CAS No.: 10022-68-1 Molecular Formula: Cd(NO3)2.4H2O Formula Weight: 308.47 (236.41anhy) Physical Form: Crystalline Density: 2.45 Melting Point: 59° Boiling Point: 132° Merck Reference: 14,1622 UN No.: 3087 MDL No.: MFCD00149626

Formule: CdN₂O₆·4H₂O
Kookpunt: 132 °C
Smeltpunt: 59 °C
Dichtheid: 2,45
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149626
CAS nummer: 10022-68-1
EINECS: 233-710-6
UN: 3087
ADR: 5.1,II
Merck Index: 14,1622

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR