Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41805.FF 41805.FI 41805.GH
41805.FFEA 189 EUR
41805.FF 41805.FI 41805.GH
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Platinum iridium alloy (80:20 w%)

Purity: 99.9% Molecular Formula: Pt:Ir; 80:20 wt% Physical Form: ≈0.34g/25x25mm Odour: Odourless MDL No.: MFCD02091683

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR