Palladium 5% on alumina, pellets 3 mm

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41825.14 41825.22 41825.06
41825.14EA 427 EUR
41825.14 41825.22 41825.06
Palladium 5% on alumina, pellets 3 mm
Palladium

CAS No.: 7440-05-3 Odour: Odourless MDL No.: MFCD03613602


Applications: Hydrogen removal "getter"

Formule: Pd
MW: 106.420 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR