Cetrimonium chloride ≥96%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

41861.06 41861.14 41861.22 41861.36
41861.06EA 39.4 EUR
41861.06 41861.14 41861.22 41861.36
Cetrimonium chloride ≥96%
Cetrimonium chloride

Purity: 96% CAS No.: 112-02-7 Molecular Formula: C19H42ClN Formula Weight: 320.00 Physical Form: Powder Density: 0.968 Appearance: Off-white Odour: Odourless Melting Point: 234° Beilstein Reference: 3657974 UN No.: 1759 MDL No.: MFCD00011773

Formule: C₁₉H₄₂ClN
Smeltpunt: 234 °C
Dichtheid: 0,968
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011773
CAS nummer: 112-02-7
EINECS: 203-928-6
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR