Aluminium ≥99% (metals basis), spheres, Ø 2.38 mm (0.094 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Aluminum

42029.KMEA 50.2 EUR
42029.KM 42029.KN
Aluminium ≥99% (metals basis), spheres, Ø 2.38 mm (0.094 in)
Aluminium

Purity: 99% CAS No.: 7429-90-5 Molecular Formula: Alloy 1100, Al:Cu; 99+:0.12 wt% MDL No.: MFCD00213972

Formule: Al
MW: 26.98 g/mol
Kookpunt: 2327
Smeltpunt: 660
Dichtheid: 2.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR