Chroom(III)chloride hexahydraat ≥99.5%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Chromium trichloride hexahydrate, Chromic chloride hexahydrate

42114.22 42114.36
42114.22EA 67.2 EUR
42114.22 42114.36
Chroom(III)chloride hexahydraat ≥99.5%
Chroom(III)chloride 6H2O

Purity: 99.5% min CAS No.: 10060-12-5 Molecular Formula: CrCl3.6H2O Formula Weight: 266.45 (158.35anhy) Physical Form: Crystalline Density: 1.76 Appearance: Dark green to black Odour: Odourless Melting Point: 83° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00149660

Formule: CrCl₃·6H₂O
MW: 266.45 g/mol
Smeltpunt: 83 °C
Dichtheid: 1.76
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149660
CAS nummer: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 13,02242

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR