Chroom ≥99.95% (metals basis), cubes typ. 6.35 mm (0.25 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42310.09 42310.18 42310.30
42310.09EA 50 EUR
42310.09 42310.18 42310.30
Chroom ≥99.95% (metals basis), cubes typ. 6.35 mm (0.25 in)
Chroom

Purity: 99.95% CAS No.: 7440-47-3 Odour: Odourless MDL No.: MFCD00010944

Formule: Cr
Kookpunt: 2469 °C (1013 hPa)
Smeltpunt: 1840 °C
Dichtheid: 7,2 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010944
CAS nummer: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR