Ijsazijn 99.7+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

423220025 423225000 423220100
423220025.EA 102 EUR
423220025. 423225000. 423220100.
Ijsazijn 99.7+% ACS
Azijnzuur
Formule: H₃CCOOH
Kookpunt: 118 °C (1013 hPa)
Dichtheid: 1,048 g/cm³
Vlampunt: 40 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036152
CAS nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
Bereik: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≤0.01 % Acetic anhydride ((CH3CO)2O)
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Dilution test Passes test
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.5 ppm
Titrable base ≤0.0004 meq/g
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR