Azijnzuuranhydride 97+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

423230010 423235000 423230025
423230010.EA 57.8 EUR
423230010. 423235000. 423230025.
Azijnzuuranhydride 97+% ACS
Azijnzuuranhydride
Formule: (CH₃CO)₂O
Kookpunt: 139,9 °C (1013 hPa)
Dichtheid: 1,087 g/cm³
Vlampunt: 54 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008705
CAS nummer: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
Bereik: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
GC ≥97.0 %
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR