(3-Chloropropyl)trimethoxysilane ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


Packaged under argon in resealable ChemSeal™ bottles.

42413.AD 42413.AK 42413.K2
42413.ADEA 29.1 EUR
42413.AD 42413.AK 42413.K2
(3-Chloropropyl)trimethoxysilane ≥97%
(3-Chloropropyl)trimethoxysilane

Packaged under Argon in resealable ChemSeal® bottles


Purity: 97+% CAS No.: 2530-87-2 Molecular Formula: ClCH2CH2CH2Si(OCH3)3 Formula Weight: 198.72 Flash Point: 78°(172°F) Physical Form: Liquid Density: 1.077 Refractive Index: 1.4200 Sensitivity: Moisture Sensitive Odour: Recognizable Melting Point: -50° Boiling Point: 195-197° Beilstein Reference: 969627 MDL No.: MFCD00000997

Formule: ClH₂CCH₂CH₂Si(OCH₃)₃
Kookpunt: 195…197 °C
Smeltpunt: –50 °C
Dichtheid: 1,077
Vlampunt: 78 °C (172 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000997
CAS nummer: 2530-87-2
EINECS: 219-787-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR