Zwavelzuur 95% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

424520025 424525000 424520026 424520100 424525001
424520025.EA 89.3 EUR
424520025. 424525000. 424520026. 424520100. 424525001.
Zwavelzuur 95% ACS
Zwavelzuur
Formule: H₂SO₄
Kookpunt: ∼335 °C (1013 hPa)
Smeltpunt: 10 °C
Dichtheid: 1,83 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Appearance Free from suspended or insoluble matter
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH 95.0 to 98.0 %
Chloride (Cl) ≤0.2 ppm
Nitrate (NO3) ≤0.5 ppm
Ammonium (NH4) ≤2 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Mercury (Hg) ≤5 ppb
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤2 ppm (as SO2)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR