Ground and waste water pollution standard solution 4, Specpure®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42579.AEEA 239 EUR
42579.AE
Ground and waste water pollution standard solution 4, Specpure®
Standaarden Standaarden voor milieuanalyse

UN No.: 3264
MDL No.: MFCD03792306

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR