Zink ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42637.BU 42637.BY
42637.BUEA 114 EUR
42637.BU 42637.BY
Zink ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Zink

Purity: 99.999% CAS No.: 7440-66-6 Physical Form: ≈22.4g/m Odour: Odourless MDL No.: MFCD00011291

Formule: Zn
Kookpunt: 908 °C (1013 hPa)
Smeltpunt: 420 °C
Dichtheid: 7,14 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR