Zirconium(IV) Propoxide ∼ 70%, AcroSeal®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

427931000 427938000
427931000.EA 41.5 EUR
427931000. 427938000.
Zirconium(IV) Propoxide ∼ 70%, AcroSeal®
Zirconium(IV) Propoxide

Appearance: Colorless to yellow to amber Solution

Formule: C₁₂H₂₈O₄Zr
Dichtheid: 1,044 g/cm³
Vlampunt: 23 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00015307
CAS nummer: 23519-77-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow to amber Solution
Assay 27.0 to 29.0 % (ZrO2)
Melting point 73 to 80 °C
GC ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR