Platinum rhodium alloy (90:10 w%), draad for ISA type S standard grade thermocouple, Ø 0.203 mm (0.008 in)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42894.G1 42894.G5 42894.G9
42894.G1EA 435 EUR
42894.G1 42894.G5 42894.G9
Platinum rhodium alloy (90:10 w%), draad for ISA type S standard grade thermocouple, Ø 0.203 mm (0.008 in)
Platinum rhodium alloy (90:10 w%)
Molecular Formula: Pt:Rh; 90:10 wt% Physical Form: ≈0.65g/m Odour: Odourless MDL No.: MFCD00198186
Order Now
Specificaties
Environmentally Preferable